L(-)-Glutathione (oxidised form) ≥95%, high purity
Supplier: VWR
0524-1G
0524-5G
97061-842EA
123.72
USD
97061-842
97061-844
L(-)-Glutathione (oxidised form) ≥95%, high purity
L(-)-Glutathione (oxidized form)
Formula:
C₂₀H₃₂N₆O₁₂S₂ Melting Pt: 185 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00063106 CAS Number: 27025-41-8 EINECS: 248-170-7 |
Specification Test Results
Iron |
|
Lead |
|
Loss on Drying |
|
Purity |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...