L(-)-Glutathione (oxidised form) ≥95%, high purity

Supplier: VWR

0524-1G 0524-5G
97061-842EA 123.72 USD
97061-842 97061-844
L(-)-Glutathione (oxidised form) ≥95%, high purity
L(-)-Glutathione (oxidized form)
Formula: C₂₀H₃₂N₆O₁₂S₂
Melting Pt: 185 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063106
CAS Number: 27025-41-8
EINECS: 248-170-7

Order Now

Specification Test Results

Iron 0.002 %
Lead 0.001 %
Loss on Drying = 8 %
Purity = 95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR