Tris(hydroxymethyl)aminomethane (TRIS, Trometamol), Ultrapure

Supplier: VWR
Warning

0497-500G 0497-10KG 0497-12KG 0497-1KG 0497-50KG 0497-5KG
97061-796CS 994.77 USD
97061-796 97061-790 97061-792 97061-794 97061-798 97061-800
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol), Ultrapure
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)

Reagent for stabilization of buffer formulations and electrophoresis of biological molecules.

Formula: NH₂C(CH₂OH)₃
Boiling Pt: 288 °C
Melting Pt: 169 °C
Density: 1.328 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004679
CAS Number: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Abs.@280nm (1M, Water) 0.05
Abs.@290nm (40%, Water) = 0.2
Abs.@400nm (40%, Water) = 0.05
Arsenic = 0.0005%
Copper = 0.0001%
Insolubles = 0.005%
Iron = 0.0001%
Lead = 0.0001%
Magnesium = 0.0005%
Melting Point 168°C - 172°C
Moisture (KF) = 1%
pH (5%, Water) @25°C 10 - 11.5
Purity = 99.9%
Solubility (10%, Water) Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR