3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulfate (CHAPS), Ultrapure
Supplier: VWR
|
Warning
|
Synonyms:
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate, 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulphonate, CHAPS
0465-10G
0465-5G
0465-50G
97061-716EA
271.83
USD
97061-716
97061-720
97061-718
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulfate (CHAPS), Ultrapure
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)
Zwitterionic, both positively and negatively charged. Utilized for applications ranging from solubilizing membrane proteins in non-denaturing conditions to creating denaturing conditions during gel electrophoresis.
Formula:
C₃₂H₅₈N₂O₇S MW: 614.89 g/mol Melting Pt: 116 Storage Temperature: Refrigerator |
MDL Number:
MFCD00081083 CAS Number: 75621-03-3 Merck Index: 13,02052 |
Specification Test Results
Conductivity (10%, Water) |
|
DNase | none |
Solubility (20%, Water) | pass |
pH (10%, Water) @25°C | 5 - 7 |
Protease | none detected |
RNase | none detected |
Residue on Ignition |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...