X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) ≥99% (by enzymatic purity), Ultra Pure Grade

Supplier: VWR

Synonyms: BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

0428-100MG 0428-1G
97061-646EA 66.6 USD
97061-646 97061-648
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) ≥99% (by enzymatic purity), Ultra Pure Grade
X-Gal (5-Bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
Formula: C₁₄H₁₅BrClNO₆
MW: 408.63 g/mol
Melting Pt: 237…239 °C
Storage Temperature: Freezer
MDL Number: MFCD00005666
CAS Number: 7240-90-6
EINECS: 230-640-8

Order Now

Specification Test Results

Blank (A620; 0.1%, PO4, pH 7.3) ≤ 0.01
Identification PASS / Fail
Purity (Enzymatic) ≥ 99.0 %
Solubility (2%, DMF) PASS / Fail
Specific Rotation (0.1%, 50% DMF) -64 - -56 degrees

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR