L(+)-Glutamic acid free acid, high purity

Supplier: VWR

Synonyms: (S)-(+)-2-Aminopentanedioic acid, L(+)-1-Aminopropane-1,3-dicarboxylic acid, (S)-(+)-α-Glutamic acid, H-Glu-OH, L-Glutamic acid, L(+)-2-Aminopentanedioic acid, (S)-(+)-Glutamic acid, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid, L(+)-α-Glutamic acid

0421-1KG
97061-634CS 532.7 USD
97061-634
L(+)-Glutamic acid free acid, high purity
L(+)-Glutamic acid
Formula: HO₂CCH₂CH₂CH(NH₂)CO₂H
MW: 147.13 g/mol
Melting Pt: 205 °C
MDL Number: MFCD00002634
CAS Number: 56-86-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1cb2faefcc53483f0b3bfccd6c12b611e487241&vid=a462989da97f7186f34fef7de9cad6596c2589ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Heavy Metals (as Pb) = 0.002 %
Lead = 0.001 %
Loss on Drying = 0.1 %
Purity (Dried) 98.5 % - 101.5 %
Specific Rotation (Dried) 31.5 ° - 32.5 °

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR