Potassium chloride ACS

Supplier: VWR

Synonyms: Potassiumchloride

0395-12KG 0395-1KG 0395-2.5KG 0395-500G
97061-560EA 424.77 USD
97061-560 97061-562 97061-564 97061-566 10799-440
Potassium chloride ACS
Potassium chloride
Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1420 °C (1013 hPa)
Melting Pt: 773 °C
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Order Now

Specification Test Results

Barium Test Pass
Bromide = 0.01%
Calcium = 0.002%
Chlorate & Nitrate (as NO3) = 0.003%
Heavy Metals = 0.0005%
Insolubles = 0.005%
Iodide = 0.002%
Iron = 0.0003%
Magnesium = 0.001%
pH (5%, Water) @25°C 5.4 - 8.6
Phosphate = 0.0005%
Purity 99% - 100.5%
Sodium = 0.005%
Sulfate = 0.001%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR