Iron(II) sulfate heptahydrate ≥99% ACS

Supplier: VWR
Warning

Synonyms: Iron vitriol, Ferrous sulphate heptahydrate, Ferrous sulfate heptahydrate

0387-500G
97061-538EA 93.25 USD
97061-538 97061-540 97061-542
Iron(II) sulfate heptahydrate ≥99% ACS
Iron (II) sulphate heptahydrate
Formula: FeSO₄·7H₂O
MW: 278.02 g/mol
Melting Pt: 64 °C
Density: 1.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149719
CAS Number: 7782-63-0
EINECS: 231-753-5
Merck Index: 13,04091

Order Now

Specification Test Results

Calcium 0.005%
Chloride = 0.001%
Copper = 0.005%
Ferric Iron = 0.1%
Insolubles = 0.01%
Magnesium = 0.002%
Manganese = 0.05%
Phosphate = 0.001%
Potassium = 0.002%
Purity = 99%
Sodium = 0.02%
Zinc = 0.005%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR