Congo red, Certifiable Grade

Supplier: VWR
Danger

0379-100G
97061-520EA 131.68 USD
97061-520
Congo red, Certifiable Grade
Congo Red

pH indicator goes from blue to red over a range of 3.0 to 5.0. A cytoplasmic stain for tissues fixed in BouinÆs. Also a specific stain for amyloid.

Formula: C₃₂H₂₂N₆Na₂O₆S₂
Melting Pt: > 360 °C
Storage Temperature: Ambient
MDL Number: MFCD00004028
CAS Number: 573-58-0
EINECS: 209-358-4
Merck Index: 12,02562

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR