Magnesium sulfate, anhydrous, Reagent Grade

Supplier: VWR

Synonyms: MgSO4, Magnesium sulfate anhydrous, Magnesium sulphate anhydrous

0338-500G
97061-438CS 716.65 USD
97061-438
Magnesium sulfate, anhydrous, Reagent Grade
Magnesium sulphate
Formula: MgSO₄
MW: 120.37 g/mol
Melting Pt: 1127 °C
Density: 2.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
EINECS: 231-298-2
Merck Index: 12,05731
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Moisture 0.2 %
Purity 98 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR