Adenosine free base
Supplier: VWR
0325-100G
97061-408CS
2705.79
USD
97061-408
Adenosine free base
Adenosine
A purine nucleoside essential for a variety of metabolic processes including energy transfer, signal transduction and mediation of inflammatory pathways.
Formula:
C₁₀H₁₃N₅O₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD00005752 CAS Number: 58-61-7 EINECS: 200-389-9 |
Specification Test Results
Em (257nm, 0.1N HCl, pH 2.0) |
|
Loss on Drying |
|
Purity |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...