EDTA (ethylenediamine tetraacetic acid) free acid, Ultrapure

Supplier: VWR
Warning

0322-1KG 0322-500G
97061-404CS 314.19 USD
97061-404 97061-406
EDTA (ethylenediamine tetraacetic acid) free acid, Ultrapure
EDTA (Ethylenediamine tetraacetic acid)

Calcium chelator used to eliminate inhibition of enzyme catalyzed reactions due to traces of heavy metals.

Formula: C₁₀H₁₆N₂O₈
Boiling Pt: 614 °C (1013 hPa)
Melting Pt: 240 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003541
CAS Number: 60-00-4
EINECS: 200-449-4
REACH: 01-2119486399-18
Merck Index: 13,03546
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2ac89cc787258fff61bfc9612051bdc09d2233&vid=adf1cfd608e336342a4765f47271a739c6c17113&prid=web&clid=SAPDAM

Order Now

Specification Test Results

DNase none detected
Heavy Metals (as Pb) = 0.001%
Insolubles (5%, 1N NaOH) Pass
Iron = 0.005%
Magnesium = 0.0005%
Nitrilotriacetic Acid = 0.1%
pH (5%, Water) @25°C 2.5 - 3.5
Protease none detected
Purity = 99.5%
Residue after Ignition = 0.2%
RNase none detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR