Bromophenol blue sodium salt ACS

Supplier: VWR

0312-100G
97061-388EA 206.4 USD
97061-388
Bromophenol blue sodium salt ACS
Bromophenol blue sodium salt
Formula: C₁₉H₉Br₄NaO₅S
Storage Temperature: Ambient
MDL Number: MFCD00013793
CAS Number: 34725-61-6
EINECS: 252-170-2

Order Now

Specification Test Results

Clarity of Solution Pass
Visual Transition Interval (pH 3.0-4.6) Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR