VWR® TRIS Glycine Buffer, 10X Liquid Concentrate
Supplier: VWR
0307-4L 0307-1L 0307-5L
97061-380EA 161.16 USD
97061-380 97061-378 97061-382
VWR® TRIS Glycine Buffer, 10X Liquid Concentrate
Buffers Electrophoresis Buffers Running Buffers
A 1X working solution contains 0.025M Tris base and 0.192 M Glycine.
  • Convenient concentrations and packaging
pH (1X, Water) @25°C - 8.3 - 8.7