Ammonium carbonate, Ultrapure

Supplier: VWR
Warning

0305-1KG
97061-374CS 239.94 USD
97061-374 97061-376
Ammonium carbonate, Ultrapure
Ammonium carbonate
Formula: (NH₄)₂CO₃
Melting Pt: 58 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010890
CAS Number: 506-87-6
EINECS: 208-058-0
Merck Index: 13,00508

Order Now

Specification Test Results

Assay (NH3 30 %
Chloride 0.0005 %
Heavy Metals (as Pb) 0.0005 %
Insolubles 0.005 %
Iron 0.0005 %
Non-volatile Matter 0.01 %
Sulfur Compounds 0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR