Alcian blue

Supplier: VWR
Warning

0298-50G
97061-362EA 734.13 USD
97061-362 97061-364
Alcian blue
Alcian blue 8 GX

Em (610nm, Water)

Formula: C₅₆H₆₈Cl₄CuN₁₆S₄
Storage Temperature: Ambient
MDL Number: MFCD00010720
CAS Number: 33864-99-2
EINECS: 251-705-7

Order Now

Specification Test Results

Em (610nm, Water) 26000


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR