TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride), Ultrapure

Supplier: VWR
Warning

0234-500G 0234-12KG 0234-1KG 0234-5KG
97061-260CS 2586.41 USD
97061-260 97061-256 97061-258 97061-262
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride), Ultrapure
TRIS HCl (Tris-(hydroxymethyl) aminomethane hydrochloride)

Reagent for stabilization of buffer formulations and electrophoresis of biological molecules.

Formula: NH₂C(CH₂OH)₃·HCl
Melting Pt: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00012590
CAS Number: 1185-53-1
EINECS: 214-684-5
Merck Index: 12,09902
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cbdc9446fdb9bcfe3f1d992ff20a16726f98113&vid=3d668801395b81fa595401a752279006da8fd3cb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Calcium ≤0.0005%
Copper ≤0.0001%
DNase none detected
Insolubles ≤0.001%
Iron ≤0.001%
Lead ≤0.0005%
Magnesium ≤0.0005%
Manganese ≤0.0005%
pH (0.1M, Water) @25 °C 4.2 – 4.9
Protease none detected
Purity (Titration) ≥99.5%
RNase none detected
Zinc ≤0.0005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR