L(+)-Cysteine hydrochloride monohydrate, high purity

Supplier: VWR
Warning

Synonyms: (R)-(+)-Cysteine hydrochloride monohydrate, (R)-2-Amino-3-mercaptopropanoic acid hydrochloride hydrate, L-Cysteine HCI Monohydrate, H-Cys-OH.HCl.H2O, Cysteine Hydrochloride, Monohydrate, Cysteine Hydrochloride monohydrate

0206-500G 0206-250G
97061-206CS 3624.93 USD
97061-206 97061-204 97061-202
L(+)-Cysteine hydrochloride monohydrate, high purity
L(+)-Cysteine hydrochloride monohydrate
Formula: HSCH₂CH(NH₂)COOH·HCl·H₂O
MW: 175.64 g/mol
Melting Pt: 168…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00065606
CAS Number: 7048-04-6
EINECS: 200-157-7
Merck Index: 13,02810
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18703c57a6b5f299a5f38f5d92b333d3d65de497&vid=10395059845247050509d84df67aefcb3c98806c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Arsenic 0.00015 %
Chloride Content 19.8 % - 20.8 %
Heavy Metals (as Pb) 0.0015 %
Iron 0.003 %
Loss on Drying 8 % - 12 %
Purity 99 %
Residue on Ignition 0.4 %
Specific Rotation 5.7° - 6.8°
Sulfate 0.03 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR