Glycine for biotechnology
Supplier: VWR
Synonyms:
Aminoacetic acid, Glycocoll, H-Gly-OH
0167-10KG
0167-1KG
0167-50KG
0167-12KG
0167-5KG
97061-124EA
753.11
USD
97061-124
97061-128
97061-130
97061-126
97061-132
Glycine for biotechnology
Glycine
Formula:
NH₂CH₂COOH Boiling Pt: 181 °C (1013 hPa) Melting Pt: 240 °C Density: 1.64 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008131 CAS Number: 56-40-6 EINECS: 200-272-2 Merck Index: 13,04504 |
|
Specification Test Results
Abs.@260 nm (1M solution) | ≤0.1 |
Abs.@280 nm (1M solution) | ≤0.1 |
Chloride |
|
DNase | none detected |
Heavy Metals (as Pb) |
|
Loss on Drying |
|
pH (5%, Water) @25°C | 5.9 - 6.4 |
Protease | none detected |
Purity |
|
RNase | none detected |
Solubility (10%, Water) | Pass |
Sulfate |
|
Thin Layer Chromatography (TLC) | One Spot |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...