Saponin, Reagent Grade

Supplier: VWR

0163-100G
97061-114CS 2571.05 USD
97061-114
Saponin, Reagent Grade
Saponin
Density: 1.015…1.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00081981
CAS Number: 8047-15-2
EINECS: 232-462-6

Order Now

Specification Test Results

Ash = 10 %
Loss on Drying = 10 %
pH (20%, Water) @25°C 4.5 - 5.5
Solubility (20%, Water) Pass

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR