Adenosine 5'-diphosphate disodium salt (ADP disodium salt) dihydrate, Ultrapure for biochemistry
Supplier: VWR
|
Warning
|
0160-5G
97061-104EA
527.5
USD
97061-104
97061-106
Adenosine 5'-diphosphate disodium salt (ADP disodium salt) dihydrate, Ultrapure for biochemistry
Adenosine 5'-diphosphate disodium salt (ADP disodium salt) dihydrate
Adenosine-5"- diphosphate, disodium salt. Involved in energy and nucleic acid metabolism
Formula:
C₁₀H₁₃N₅Na₂O₁₀P₂·2H₂O MW: 471.167 g/mol (anhy) Storage Temperature: Freezer |
MDL Number:
MFCD00066635 CAS Number: 16178-48-6 EINECS: 240-314-7 |
Specification Test Results
A250/A260 | 0.74 - 0.82 |
A280/A260 | 0.13 - 0.19 |
Arsenic |
|
ATP |
|
Em (259nm,Phosphate Buffer,pH 7.0) |
|
Heavy Metals |
|
Loss on Drying |
|
Solubility (5%, Water) | pass |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...