L(+)-Ascorbic acid free acid ACS
Supplier: VWR
Synonyms:
Antiscorbic vitamin, Ascorbic Acid, L-Ascorbic acid, Vitamin C, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
0143-100G
0143-500G
97061-072CS
439.84
USD
97061-072
97061-074
80760-014
L(+)-Ascorbic acid free acid ACS
L(+)-Ascorbic acid
Formula:
C₆H₈O₆ MW: 176.13 g/mol Boiling Pt: 553 Melting Pt: 190…192 °C Density: 1.694 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064328 CAS Number: 50-81-7 EINECS: 200-066-2 REACH: exempt Merck Index: 13,00837 |
|
Specification Test Results
Heavy Metals (as Pb) |
|
Iron |
|
Purity |
|
Residue on Ignition |
|
Specific Rotation | 20.5° - 21.5° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...