Magnesium acetate tetrahydrate

Supplier: VWR

Synonyms: Acetic acid magnesium salt tetrahydrate, Magnesium di(acetate) tetrahydrate

0131-500G
97061-056CS 269.66 USD
97061-056 97061-060
Magnesium acetate tetrahydrate
Magnesium acetate tetrahydrate
Formula: (H₃CCOO)₂Mg·4H₂O
MW: 214.46 g/mol
Melting Pt: 80 °C
Density: 1.454 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149214
CAS Number: 16674-78-5
EINECS: 205-554-9
Merck Index: 13,05676

Order Now

Specification Test Results

Chloride = 0.01 %
Insolubles = 0.005 %
Iron = 0.01 %
Purity
Sulfate = 0.05 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR