Eosin Y (yellowish), Certifiable Grade

Supplier: VWR
Warning

0109-100G
97061-034EA 92.6 USD
97061-034
Eosin Y (yellowish), Certifiable Grade
Eosin Y (yellowish)

Counterstain to hematoxylin. Used for staining exfoliative cytology.

Formula: C₂₀H₆Br₄Na₂O₅
Melting Pt: 295…296 °C
Storage Temperature: Ambient
MDL Number: MFCD00005040
CAS Number: 17372-87-1
EINECS: 241-409-6
Merck Index: 13,03634

Order Now

Specification Test Results

Dye Content 88%
Lambda Max 515 – 518 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR