Ammonium acetate, Ultrapure

Supplier: VWR
Warning

0103-500G 0103-2.5KG
97061-014CS 488.36 USD
97061-014 97061-012
Ammonium acetate, Ultrapure
Ammonium acetate
Formula: H₃CCOONH₄
Melting Pt: 114 °C
Density: 1.17 g/cm³ (20 °C)
Flash Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD00013066
CAS Number: 631-61-8
EINECS: 211-162-9
Merck Index: 13,00495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Abs.@280nm (1M, Water) 0.01 %
Abs.@354nm (1M, Water) 0.01 %
Chloride 0.0005 %
Heavy Metals (as Pb) 0.005 %
Insolubles 0.0005 %
Iron 0.001 %
Nitrate 0.001 %
pH (5%, Water) @25°C 6.7 - 7.3
Purity 97.0 %
Residue after Ignition 0.01 %
Sulfate 0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR