Water, Reagent Grade, meets analytical specification of USP, sterile, Corning®

Supplier: Corning

46-002-LF
97060-878PK 89.23 USD
97060-878
Water, Reagent Grade, meets analytical specification of USP, sterile, Corning®
Water

Reagent grade water is ideal for the preparation of reagents, rinsing glassware and plastic ware, cleaning electrodes and other common laboratory tasks requiring highly pure water.


  • No toxic agents, including DEPC, are used in its preparation
  • RNAse, DNAse and Protease free


The design allows for portability, ease of storage and convenient dispensing. Upon packaging, the water meets USP conductivity requirements for water for injection (WFI) and is RNAse-, DNAse- and Protease-free. Provided in packs of 2 x 4L cubes.

Formula: H₂O
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR