Incubator-Genie™ Incubator Shaker Rotator Rocker and Hybridization Oven, 230 V
Supplier: Scientific Industries
SI-1401
97060-140EA 7464.75 USD
97060-140
Incubator-Genie™ Incubator Shaker Rotator Rocker and Hybridization Oven, 230 V
Shakers and Mixers Shaking Incubators
Multi-functional incubator shaker and hybridization oven designed to incubate, rotate, rock, and shake all in one compact unit.
  • 230 V model
  • Compact unit
  • Multi-functional
Temperature control from 28 °C to 75 °C, magnetic stainless steel platform allows for easily mounting containers with magnetic clips or strips, and may be set to rock or rotate almost any container. A variety of accessories are available to provide maximum application versatility, including hybridization tubes.
Certifications: CE marked.
Ordering information: Magnetic platform,More Product Information