Incubator-Genie™ Incubator Shaker Rotator Rocker and Hybridization Oven, Scientific Industries
Supplier: Scientific Industries
SI-1400
97060-138EA 5687.16 USD
97060-138
Incubator-Genie™ Incubator Shaker Rotator Rocker and Hybridization Oven, Scientific Industries
Shakers and Mixers Shaking Incubators
Multi-functional incubator shaker and hybridization oven designed to Incubate, Rotate, Rock, and Shake all in one compact unit.
  • Compact unit
  • Multi-functional
Temperature control from 28°C to 75°C, magnetic stainless steel platform allows for easily mounting containers with magnetic clips or strips, and may be set to rock or rotate almost any container. A variety of accessories are available to provide maximum application versatility, including hybridization tubes.
Certifications: CE marked.
Ordering information: Magnetic Platform, 1 Clip Plate for each 10-13mm, 15-17mm, and 28-30mm tubes, 4 Magnetic Bag Mounting Strips, Non-Skid Tray, and Stackable Shelf included. The 230V model is supplied without a plug.