Polytron® Generator PT-DA 03/2EC-050, Kinematica
Supplier: Kinematica
11030004
97057-704EA 2151.2 USD
97057-704
Polytron® Generator PT-DA 03/2EC-050, Kinematica
Homogenizers Dispersing Tools
The smallest available generator in the industry for samples as small as 100 microliters.
  • Easy clean design for quick disasembly
  • 316L SS
  • Fits in micro-centrifuge tubes
When only the smallest tissue of cell samples to be homogenized, our industry exclusive 3 mm design allows for samples as small as 100 microliters.