Polytron® Homogenizer PT 2500E, Kinematica
Supplier: Kinematica
11010055
97057-702EA 3057.93 USD
97057-702
Polytron® Homogenizer PT 2500E, Kinematica
Homogenizers Benchtop Homogenizers
Desinged by benchtop operation, this versatile unit accepts the same generators as our handheld units, as well as several larger sizes for bigger sample volumes.
  • Digital speed indicator allows for easy control and repilcation of operating parameters
  • Quick connect coupling for swapping out generators
  • Rugged contrucion with easy to clean housing
This verastile unit accepts our 3, 5, 7, 12, 20, and 25 mm generators for sample sizes between 0.01 mL and 2000 mL. Specialty generators are available with designs for fibrous, brittle, and highly viscous samples. The powerful 500 W motor 30000 rpm motor provides ample power for even viscous samples.