Hex-Type Valve Wrench, MicroSolv Technology™
Supplier: MicroSolv Technology Corp
83501-30
97055-226EA 19.87 USD
97055-226
Hex-Type Valve Wrench, MicroSolv Technology™
Wrenches
This hex-type valve wrench is for columns with an ID of 1.5, 2.0, 3.0mm.