Spectronic 200 Visible Spectrophotometer, Thermo Scientific
Supplier: Thermo Scientific
840-281700
97054-250EA 3173 USD
97054-250
Spectronic 200 Visible Spectrophotometer, Thermo Scientific
Spectrophotometers and Photometers UV/Visible Spectrophotometers
SPECTRONIC 200 is a modern, visible spectrophotometer for education and routine industrial use
USB connectivity provides additional flexibility for data and method storage, printing, or connection to a PC with VISIONlite 5 (or later) software for more involved experiments, data processing and storage.
Built upon a foundation of over 50 years of experience the SPECTRONIC 200 features everything your existing visible spectrophotometer offers and more. Familiar controls, a full color display, and built-in software provides a full range of functionality including live display mode, full spectrum measurements, multi-wavelength analysis, and quantitative analysis with up to 4 standards. A programmable analyzer mode allows you to streamline repetitive procedures.
Ordering information: One-year manufacturer’s warranty. Sample compartment accepts standard 10-mm square cuvettes and test tube cuvettes from 13- to 2-mm diameter.