Oxygen Pen, Sper Scientific
Supplier: Sper Scientific
800047
97052-182EA 1195.46 USD
97052-182
Oxygen Pen, Sper Scientific
Electrochemistry Meters Oxygen Meters
Reads oxygen in air for environmental studies, IAQ, bio-technology (for oxygen incubators and anaerobic cultivators), food storage and refrigeration, and testing fire alarm and fuel cell systems
The fast, highly reliable, galvanic oxygen sensor is unaffected by acidic gases such as CO2. Simultaneously displays O2 concentration and air temperature in °C or °F. Features hold, min/max, a low battery indicator, ATC and auto-power-off.
Ordering information: 1-year warranty. Comes with 4 AAA batteries and a soft carrying case with a belt loop and shoulder strap.