AcroPrep™ Advance 96-Well Filter Plates, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
8049
97052-118PK 241.92 USD
97052-118
AcroPrep™ Advance 96-Well Filter Plates, Cytiva (Formerly Pall Lab)
Microplates Filter Plates
AcroPrep™ Advance Filter Plates feature polypropylene plate housing, polystyrene lid, and borosilicate glass filter media.
  • Polypropylene housing
  • Polystyrene lid
  • Individually sealed wells
Individually sealed membranes and biologically inert housing minimizes the possibility of sample cross-contamination or loss. New sealing technology minimizes sample weeping during incubations. Designed to meet SBS/ANSI guidelines, ensuring compatibility with standard processing equipment. Smooth top surface, textured window for labeling, and A1 corner notch allow for easy identification.
Well bottom area: 0.25 cm²
Dimensions: 12.78 x 8.55 x 1.44 cm (5.03 x 3.36 x 0.56")