Screw Caps with Partial Slit Septa for 12×32 mm Vials, Microsolv Technology Corp
Supplier: MicroSolv Technology Corp
9502S-34C-B
97051-404PK 78.5 USD
97051-404
Screw Caps with Partial Slit Septa for 12×32 mm Vials, Microsolv Technology Corp
Closures Test/Sample Tube Closures
Screw Caps have a partial precut Y shape slit to prevent evaporation of volatile samples before injection
Helps to minimize vacuum forming in the vial.