Beta Tool™, Tubing Straightener, MicroSolv
Supplier: MicroSolv Technology Corp
46010-BT
97051-378EA 215.23 USD
97051-378
Beta Tool™, Tubing Straightener, MicroSolv
Pliers
Beta Tool™ is a pair of precision, high grade pliers and can be used as needle nose pliers or as a tubing straightener.
  • Use to remove kinks or bends in standard outer diameter HPLC stainless steel tubing
It brings 1/16" outer dia. stainless steel tubing back to a perfect round without compromising the inner diameter.