DigiTUBE® Digestion Tubes
Supplier: PerkinElmer
N9308008 N9308016 N9308037 N9308066
97050-066PK 894.84 USD
97050-066 97050-082 97050-124 97050-182
DigiTUBE® Digestion Tubes
Tubes Digestion Tubes
Disposable sample digestion tubes with Class A graduations.
  • Easy-to-read graduations
  • Allows normalization directly in the tube
  • Easy one-handed closure of leak-free screw cap
All-in-one tubes eliminate the need for five separate vessels such as Digestion beakers, volumetric flasks, graduated cylinders, autosampler tubes and storage containers.