Cryo Marker Pen Set, Thermo Scientific
Supplier: Thermo Fisher Scientific
4000222
97049-678EA 55.46 USD
97049-678
Cryo Marker Pen Set, Thermo Scientific
Pens Cryo Markers
For fast-drying, non-smearing labeling
Recommended for all cryogenic vials at temperatures 200°C (-328°F)