Preparative TLC Plates, Agela Technologies
Supplier: Phenomenex
TZ-CSF200200-0
97049-368PK 218.57 USD
97049-368 97049-370
Preparative TLC Plates, Agela Technologies
TLC Plates
These thin layer chromatography plates are used for a variety of chemistries
They are flexible and glass-backed for easy use. When matching with Claricep flash cartridges, the TLC plates serve as a tool for reaction monitoring and method development based on Rf values for flash chromatography.