Air Jet Sieve AS 200, Verder Scientific
Supplier: Retsch
300270001
97049-034EA 13730.74 USD
97049-034
Air Jet Sieve AS 200, Verder Scientific
Shakers and Mixers Sieve Shakers
Air Jet Sieve AS 200 jet is particularly suitable for sieve cuts of powdered materials which require efficient dispersion and deagglomeration.
  • Easy operation, ergonomic design
  • Additional Quick Start mode
  • Open mesh function to reduce the number of near-mesh particles
  • Memory for 9 Standard Operating Procedures
  • Two fractions can be collected by using cyclone
  • Digital adjustment of all process parameters (time, vacuum, speed)
AS 200 jet is equipped with a rotating slotted nozzle. The sieve and lid are placed above the nozzle. A vacuum generates a strong jet of air which disperses the particles on the sieve through the slotted nozzle. The material which is smaller than the mesh size of the sieve is transported via the air flow to the cyclone or to the vacuum cleaner. The air jet helps to deagglomerate the particles and constantly purges the sieve mesh.