IKAMAG® Color Squid Magnetic Stirrer, IKA® Works
Supplier: IKA Works
0003671000
97048-750EA 1207.55 USD
97048-750
IKAMAG® Color Squid Magnetic Stirrer, IKA® Works
Stirrers Magnetic Stirrers
This improved, compact magnetic stirrer now features a new design
An LED digital speed display and an electronically controlled motor for increased capacity are two of many new features. The unit also provides outstanding chemical resistance due to a glass top and a synthetic bottom made of TPC-ET.