WHEATON® MicroLiter 9 mm Screw-Thread Vials, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
09-1000 09-1000-100 09-1000-140 09-1000-S 09-1200 09-1200-100 09-1200-101 09-1200-300 09-1300-100 09-1300-1000 09-1400 09-2200 09-2300-1000 09-3200 76-1200 76-1200-100 76-1200-101 76-1200-300 76-1200-S 76-2200 76-2200-101 76-2200-300 76-2200-S 09-1200-102 09-2200-102
97046-310PK 41.52 USD
97046-310 97046-312 97046-314 97046-316 97046-318 97046-320 97046-494 97046-496 97046-498 97046-500 97046-502 97046-676 97046-678 97046-680 97046-682 97046-746 97046-748 97046-750 97046-752 97046-926 97046-934 97048-228 97048-230 97048-232 97048-234 97048-236 97048-410 97048-412 97048-414 97048-415 89496-144 89496-146 10039-382
WHEATON® MicroLiter 9 mm Screw-Thread Vials, DWK Life Sciences
Vials
All vials may appear to be created equal, but quality and performance make the difference when analyzing vital samples. MicroLiter vials are identical, clean, and dependable, and are certified to ensure optimal performance and reproducible results.
  • Performance equal to an 11 mm crimp-top vial
  • Inserts available
MicroLiter vials are cleaned and packaged in a certified Class 10000 clean room according to a designed process, developed to eliminate chromatographic errors associated with vials
The vials and inserts are made from Type 1 borosilicate glass or polypropylene, and meet strict tolerances. Their performance is equal to an 11 mm crimp-top vial, with the convenience and accessibility of screw threads. The µl Vials™ feature conical bottoms. The 76-Series Vials™ feature a modified neck finish to ensure a consistent, repeatable seating of the gripper against the bottom of the cap.
Critical physical dimensions are fully tested, as well as chemical cleanliness testing to ensure that trace levels of residues are not present.
Certifications: Certified Clean