Economical Stainless Steel Hook Weight Set, Troemner
Supplier: Troemner
30390775
97044-290EA 308.94 USD
97044-290
Economical Stainless Steel Hook Weight Set, Troemner
Weights Calibration Weights
The makeup of Economical Stainless Steel Weight Sets varies
Hook Weights range from 10 g to 1 kg (0.1 N to 10 N) and are made of stainless steel. On all Hook Weights, except the 10 g, there is a hook on the top and the bottom of the weight so the weights can be hooked together. The 10 g (0.1 N) Hook Weight has a hook on the top only. The design of the Hook Weight allows the bottom hook to be recessed in the bottom, so the hooked weights can be set on a flat surface without tipping over. The opening of the hook for each weight is 1/2 inch.
Weights are adjusted to tolerances specified by ANSI/ASTM E617 Class 7.