VWR® Ultrasonic Cleaners
Supplier: VWR International
97043-932 97043-936 97043-940 97043-944 97043-948 97043-952 97043-956 97043-960 97043-964 97043-968 97043-972 97043-976 97043-980 97043-984 97043-988 97043-992 97043-996 97044-002 97044-006 97044-010 97044-014
97043-932EA 670.89 USD
97043-932 97043-936 97043-940 97043-944 97043-948 97043-952 97043-956 97043-960 97043-964 97043-968 97043-972 97043-976 97043-980 97043-984 97043-988 97043-992 97043-996 97044-002 97044-006 97044-010 97044-014
VWR® Ultrasonic Cleaners
Ultrasonic Cleaners
Ultrasonic cleaners provide highly effective and efficient cleaning of laboratory glassware and instruments.
  • Highly effective and efficient cleaning
  • Digital timer
  • Optional features and accessories available
All models feature a digital timer. Optional features include a heater and degassing function. Accessory baskets, covers, and support racks are constructed of Type 304 stainless steel.
Certifications: UL and C-UL listed.
Ordering information: Includes built-in drains for models 9.5L and above, and all models include a support rack and cover.