VWR® Advanced Heavy-Duty Vortex Mixers, 230V (Export Only)
Supplier: VWR International
97043-568
97043-568EA 1406.68 USD
97043-568
VWR® Advanced Heavy-Duty Vortex Mixers, 230V (Export Only)
Shakers and Mixers Vortex Mixers
Ideal for applications that demand repeatable results, mixer features touchpad controls and LED displays for speed and time.
  • 230 V model
  • Designed for continuous duty
  • LED displays for time and speed
  • Now with best-in-class, exclusive five year warranty
The heavy-duty design and efficient motor allow the mixer to operate in continuous duty and handle all accessories over the entire speed range
Microprocessor control maintains set speed for strong, consistent mixing action. Timer will display elapsed time or, when programmed to user defined limits, the unit will shut off when time reaches zero.
Choose from two modes of operation: 'Touch' mode for mixing tubes when cup head or Universal Holder with Cover is depressed or "On" mode when using any of the accessory attachments for continuous operation.
Certifications: RoHS certified. TUV listed. 230V units are CE Marked.
Ordering information: Mixer includes a cup head, Universal Holder with Cover, Foam microtube insert for (38) 1.5 to 2.0 mL microtubes and a 92" (234 cm) Euro type plug.
Caution: 230 V model.