VWR® Standard Heavy-Duty Vortex Mixer, 230 V
Supplier: VWR International
80862956
97043-566EA 1244.22 USD
97043-566
VWR® Standard Heavy-Duty Vortex Mixer, 230 V
Shakers and Mixers Vortex Mixers
Variable speed analog mixer is designed for continuous duty.
  • 230 V model
  • Multiple operation modes
  • Designed for continuous duty
  • Best-in-Class, 5-year warranty
The heavy-duty design and efficient motor allowthe mixer to operate in continuous duty and handle all accessories over the entire speed range. Choose from two modes of operation: 'Touch' mode for mixing tubes when cup head or universal holder with cover is depressed or 'On' mode when using any of the accessory attachments for continuous operation.
Certifications: RoHS certified. 230 V units are CE Marked.
Ordering information: Mixer includes a cup head, universal holder with cover, foam microtube insert for (38) 1.5 to 2.0 ml microtubes and a 92" (234 cm) Euro type plug.
Caution: 230 V model.