VWR® Illuminated Tissue Flotation Baths, 230 V
Supplier: VWR International
97043-532 97043-536
97043-532EA 2313.35 USD
97043-532 97043-536
VWR® Illuminated Tissue Flotation Baths, 230 V
Baths Tissue Baths
The VWR® Illuminated Tissue Flotation Baths are designed for use in the Histology Lab that is conducting tissue preparation.
  • 230 V models, not for use in USA
  • LED illuminated black background
  • Easy removal of sample for cleaning
  • Compact footprint
  • Easy-to-read display
An LED illuminated black background with a transparent PYREX® dish allows for easy viewing of the sample which can also be removed for easy cleaning
A compact footprint uses less bench space and the easy-to-read display can be seen easily through out the lab.
The digital microprocessor control and temperature sensor ensures temperature uniformity while the safety over-temp device provides protection for of the samples. VWR® offers both a Premium version and a Standard version of these Tissue Flotation Baths. The Premium version includes both an Orienter Block and a Dryer Block on the back of the unit which assists in the drying and smoothing of slides and specimens.
Certifications: UL/CSA Listed and CE Certified.
Ordering information: The VWR® Illuminated Tissue Flotation Bath is supplied with one PYREX® Dish, Instruction Manual and Power Cord. Each unit comes with a two-year warranty on parts and labor. Additional PYREX® dishes are available for purchase.
Caution: 230 V models, not for use in USA. Corresponding models with voltage suitable for use in US (97043-530/97043-534) are also available, please see separate product page.