ProfiLine™ Single-Parameter Portable Meter Kits, Conductivity, WTW
Supplier: WTW
2CA101
97043-142EA 2809.14 USD
97043-142 97043-228
ProfiLine™ Single-Parameter Portable Meter Kits, Conductivity, WTW
Electrochemistry Meters Conductivity Meters
Meters provide reliable measurements for field applications
The robust Model Cond 3110 meter provides easy operation in the field. The versatile Model Cond 3210 meter features a backlit graphic LCD, automatic stability control (AutoRead), an internal clock, and memory that stores up to 200 data sets.
Equipped with built-in calibration indicator and waterproof to IP67 standards.
Ordering information: Kits include a four-electrode conductivity cell TetraCon® 325, standard solution (1413 µS/cm at 25°C), carry case, stand, beaker, short instruction manual, CD-ROM, and batteries. Kit 97043-142 includes a 1m (3.3') electrode cable. Kit 97043-228 includes a 3m (9.8') electrode cable.