Electrolyte Solution, L2114, SI Analytics
Supplier: YSI
285138349
97042-458EA 112.05 USD
97042-458
Electrolyte Solution, L2114, SI Analytics
Electrolytes Electrode Filling Solutions
This solution is made up of 2 mol/l KNO3 and 0.001 mol/l KCl solution.
Packaging: Packed in 250 ml DURAN® bottle.