IR Thermometer/LED Light Pen, Sper Scientific
Supplier: Sper Scientific
800100
97035-828EA 189.16 USD
97035-828
IR Thermometer/LED Light Pen, Sper Scientific
Thermometers Digital Thermometers
One end of this metal pen features a super bright white LED. A standard AAA provides 6 hours of continuous illumination. The other end contains a non-contact Infrared thermometer.
  • Two button cell batteries provide 35 hours of continuous temperature measurement
  • Backlit LCD display will inform you when batteries need to be changed
Press the button to scan temperature and release to hold. The IR thermometer has a response time of 1 second and is protected by a lens cap when not in use.
Ordering information: 5-year warranty.
Delivery information: All batteries are included (one AAA and two L1154 Button Cell). Comes in a sturdy foam lined metal case.